N-Cyclohexyl-N-methyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052493-500MG 052493-2.5G
101899-858EA 364.62 USD
101899-858 101899-860
N-Cyclohexyl-N-methyl-4-piperidinecarboxamide hydrochloride
N-Cyclohexyl-N-methyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O
MDL Number: MFCD13562230
CAS Number: 1220033-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR