(4-Hydroxy-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052491-500MG
052491-2.5G
101899-850EA
364.62
USD
101899-850
101899-852
(4-Hydroxy-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
(4-Hydroxy-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₁ClN₂O₂ MW: 248.76 g/mol |
MDL Number:
MFCD13562228 CAS Number: 1220039-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...