4-(4-Piperidinylcarbonyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052490-500MG 052490-2.5G
101899-846EA 364.62 USD
101899-846 101899-848
4-(4-Piperidinylcarbonyl)-2-piperazinone hydrochloride
4-(4-Piperidinylcarbonyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClN₃O₂
MDL Number: MFCD13562227
CAS Number: 1220018-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR