(4-Benzyl-1-piperazinyl)(4-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052489-500MG 052489-2.5G
101899-842EA 364.62 USD
101899-842 101899-844
(4-Benzyl-1-piperazinyl)(4-piperidinyl)methanone hydrochloride
(4-Benzyl-1-piperazinyl)(4-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₆ClN₃O
MDL Number: MFCD13562226
CAS Number: 1220034-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR