(4-Methyl-1-piperazinyl)(4-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052488-500MG 052488-2.5G
101899-838EA 364.62 USD
101899-838 101899-840
(4-Methyl-1-piperazinyl)(4-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperazinyl)(4-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClN₃O
MDL Number: MFCD09972278
CAS Number: 1018826-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR