(4-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
Synonyms:
4-Methyl-1-(piperidin-4-ylcarbonyl)piperidine hydrochloride
052487-500MG
052487-2.5G
101899-834EA
364.62
USD
101899-834
101899-836
(4-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₃ClN₂O MW: 246.78 g/mol |
MDL Number:
MFCD06407989 CAS Number: 690632-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...