(3-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052486-500MG 052486-2.5G
101899-830EA 364.62 USD
101899-830 101899-832
(3-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride
(3-Methyl-1-piperidinyl)(4-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MW: 246.78 g/mol
MDL Number: MFCD11099597
CAS Number: 1170118-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR