N,N-Diallyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052484-500MG 052484-2.5G
101899-822EA 364.62 USD
101899-822 101899-824
N,N-Diallyl-4-piperidinecarboxamide hydrochloride
N,N-Diallyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₁ClN₂O
MDL Number: MFCD13562224
CAS Number: 1220039-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR