(3-Hydroxy-1-pyrrolidinyl)(4-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052481-500MG 052481-2.5G
101899-810EA 364.62 USD
101899-810 101899-812
(3-Hydroxy-1-pyrrolidinyl)(4-piperidinyl)methanone hydrochloride
(3-Hydroxy-1-pyrrolidinyl)(4-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O₂
MW: 234.73 g/mol
MDL Number: MFCD13562222
CAS Number: 1220039-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR