1-Piperidinyl(4-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052480-500MG
052480-2.5G
101899-806EA
364.62
USD
101899-806
101899-808
1-Piperidinyl(4-piperidinyl)methanone hydrochloride
1-Piperidinyl(4-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₁ClN₂O |
MDL Number:
MFCD01580851 CAS Number: 63214-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...