N-(2-Hydroxyethyl)-N-methyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052479-500MG 052479-2.5G
101899-802EA 364.62 USD
101899-802 101899-804
N-(2-Hydroxyethyl)-N-methyl-4-piperidinecarboxamide hydrochloride
N-(2-Hydroxyethyl)-N-methyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562221
CAS Number: 1220033-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR