N,N-Diethyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052478-500MG 052478-2.5G
101899-798EA 364.62 USD
101899-798 101899-800
N,N-Diethyl-4-piperidinecarboxamide hydrochloride
N,N-Diethyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD11615738
CAS Number: 95389-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR