N,N-Dimethyl-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052476-500MG
052476-2.5G
101899-790EA
336.92
USD
101899-790
101899-792
N,N-Dimethyl-4-piperidinecarboxamide hydrochloride
N,N-Dimethyl-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O |
MDL Number:
MFCD07781032 CAS Number: 6270-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...