N-Benzyl-N-ethyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052474-500MG 052474-2.5G
101899-782EA 364.62 USD
101899-782 101899-784
N-Benzyl-N-ethyl-3-piperidinecarboxamide hydrochloride
N-Benzyl-N-ethyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O
MDL Number: MFCD13562219
CAS Number: 1220033-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR