3,4-Dihydro-1(2H)-quinolinyl(3-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052473-500MG 052473-2.5G
101899-778EA 364.62 USD
101899-778 101899-780
3,4-Dihydro-1(2H)-quinolinyl(3-piperidinyl)methanone hydrochloride
3,4-Dihydro-1(2H)-quinolinyl(3-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₁ClN₂O
MDL Number: MFCD13562218
CAS Number: 1220034-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR