N-Cyclohexyl-N-ethyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052469-500MG 052469-2.5G
101899-762EA 364.62 USD
101899-762 101899-764
N-Cyclohexyl-N-ethyl-3-piperidinecarboxamide hydrochloride
N-Cyclohexyl-N-ethyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₇ClN₂O
MDL Number: MFCD13562214
CAS Number: 1219972-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR