N-Benzyl-N-methyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052468-500MG 052468-2.5G
101899-758EA 364.62 USD
101899-758 101899-760
N-Benzyl-N-methyl-3-piperidinecarboxamide hydrochloride
N-Benzyl-N-methyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O
MDL Number: MFCD13562213
CAS Number: 1220039-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR