(2-Ethyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052467-500MG
052467-2.5G
101899-754EA
364.62
USD
101899-754
101899-756
(2-Ethyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
(2-Ethyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₅ClN₂O MW: 260.81 g/mol |
MDL Number:
MFCD13562212 CAS Number: 1220033-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...