N-Cyclohexyl-N-methyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052466-500MG 052466-2.5G
101899-750EA 364.62 USD
101899-750 101899-752
N-Cyclohexyl-N-methyl-3-piperidinecarboxamide hydrochloride
N-Cyclohexyl-N-methyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O
MDL Number: MFCD13562211
CAS Number: 1220033-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR