(4-Hydroxy-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
(4-Hydroxypiperidin-1-yl)piperidin-3-ylmethanone
052464-500MG
052464-2.5G
101899-742EA
258.46
USD
101899-742
101899-744
(4-Hydroxy-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
(4-Hydroxy-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
CAS: 496057-57-9
MDL Number: MFCD06796403
Molecular Formula: C11H21ClN2O2
Molecular Weight: 248,76
Hazard: Irritant
Formula:
C₁₁H₂₁ClN₂O₂ MW: 248.76 g/mol |
MDL Number:
MFCD06796403 CAS Number: 496057-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...