4-(3-Piperidinylcarbonyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052463-500MG 052463-2.5G
101899-738EA 364.62 USD
101899-738 101899-740
4-(3-Piperidinylcarbonyl)-2-piperazinone hydrochloride
4-(3-Piperidinylcarbonyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClN₃O₂
MDL Number: MFCD13562209
CAS Number: 1220033-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR