1-Azepanyl(3-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052462-500MG 052462-2.5G
101899-734EA 364.62 USD
101899-734 101899-736
1-Azepanyl(3-piperidinyl)methanone hydrochloride
1-Azepanyl(3-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MDL Number: MFCD13562208
CAS Number: 1220039-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR