(4-Benzyl-1-piperazinyl)(3-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052461-500MG
052461-2.5G
101899-730EA
364.62
USD
101899-730
101899-732
(4-Benzyl-1-piperazinyl)(3-piperidinyl)methanone hydrochloride
(4-Benzyl-1-piperazinyl)(3-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₆ClN₃O MW: 323.87 g/mol |
MDL Number:
MFCD13562207 CAS Number: 1220031-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...