(4-Methyl-1-piperazinyl)(3-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052460-500MG 052460-2.5G
101899-726EA 364.62 USD
101899-726 101899-728
(4-Methyl-1-piperazinyl)(3-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperazinyl)(3-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClN₃O
MW: 247.77 g/mol
MDL Number: MFCD13562206
CAS Number: 1217120-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR