(2-Methyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052457-500MG
052457-2.5G
101899-714EA
364.62
USD
101899-714
101899-716
(2-Methyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
(2-Methyl-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₃ClN₂O |
MDL Number:
MFCD13562204 CAS Number: 1220033-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...