N-Ethyl-N-(2-hydroxyethyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052455-500MG 052455-2.5G
101899-706EA 364.62 USD
101899-706 101899-708
N-Ethyl-N-(2-hydroxyethyl)-3-piperidinecarboxamide hydrochloride
N-Ethyl-N-(2-hydroxyethyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD13562202
CAS Number: 1220028-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR