N-Butyl-N-methyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052454-500MG 052454-2.5G
101899-702EA 364.62 USD
101899-702 101899-704
N-Butyl-N-methyl-3-piperidinecarboxamide hydrochloride
N-Butyl-N-methyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD13562201
CAS Number: 937725-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR