(3-Hydroxy-1-pyrrolidinyl)(3-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052453-500MG
052453-2.5G
101899-698EA
364.62
USD
101899-698
101899-700
(3-Hydroxy-1-pyrrolidinyl)(3-piperidinyl)methanone hydrochloride
(3-Hydroxy-1-pyrrolidinyl)(3-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₉ClN₂O₂ MW: 234.73 g/mol |
MDL Number:
MFCD13562200 CAS Number: 1220028-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...