4-Morpholinyl(3-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052452-500MG 052452-2.5G
101899-694EA 364.62 USD
101899-694 101899-696
4-Morpholinyl(3-piperidinyl)methanone hydrochloride
4-Morpholinyl(3-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O₂
MW: 234.73 g/mol
MDL Number: MFCD07781104
CAS Number: 35090-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR