N-(2-Hydroxyethyl)-N-methyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052450-500MG 052450-2.5G
101899-686EA 364.62 USD
101899-686 101899-688
N-(2-Hydroxyethyl)-N-methyl-3-piperidinecarboxamide hydrochloride
N-(2-Hydroxyethyl)-N-methyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562199
CAS Number: 1220028-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR