3-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052448-500MG 052448-2.5G
101899-678EA 364.62 USD
101899-678 101899-680
3-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride
3-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O
MW: 218.73 g/mol
MDL Number: MFCD11520873
CAS Number: 937724-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR