N,N-Dimethyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052447-500MG 052447-2.5G
101899-674EA 364.62 USD
101899-674 101899-676
N,N-Dimethyl-3-piperidinecarboxamide hydrochloride
N,N-Dimethyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD07781029
CAS Number: 112950-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR