N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052446-500MG 052446-2.5G
101899-670EA 364.62 USD
101899-670 101899-672
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-piperidinecarboxamide hydrochloride
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O₂
MDL Number: MFCD13562198
CAS Number: 1246172-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR