N-Benzyl-N-ethyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052445-500MG 052445-2.5G
101899-666EA 364.62 USD
101899-666 101899-668
N-Benzyl-N-ethyl-2-piperidinecarboxamide hydrochloride
N-Benzyl-N-ethyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O
MDL Number: MFCD13562197
CAS Number: 1246172-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR