3,4-Dihydro-1(2H)-quinolinyl(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052444-500MG 052444-2.5G
101899-662EA 364.62 USD
101899-662 101899-664
3,4-Dihydro-1(2H)-quinolinyl(2-piperidinyl)methanone hydrochloride
3,4-Dihydro-1(2H)-quinolinyl(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₁ClN₂O
MW: 280.8 g/mol
MDL Number: MFCD13562196
CAS Number: 1236254-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR