2,3-Dihydro-1H-indol-1-yl-(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052443-500MG 052443-2.5G
101899-658EA 364.62 USD
101899-658 101899-660
2,3-Dihydro-1H-indol-1-yl-(2-piperidinyl)methanone hydrochloride
2,3-Dihydro-1H-indol-1-yl-(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O
MDL Number: MFCD13562195
CAS Number: 1236256-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR