[4-(2-Hydroxyethyl)-1-piperazinyl](2-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052441-500MG
052441-2.5G
101899-650EA
364.62
USD
101899-650
101899-652
[4-(2-Hydroxyethyl)-1-piperazinyl](2-piperidinyl)methanone hydrochloride
[4-(2-Hydroxyethyl)-1-piperazinyl](2-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₄ClN₃O₂ MW: 277.8 g/mol |
MDL Number:
MFCD13562193 CAS Number: 1246172-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...