N-Cyclohexyl-N-ethyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052440-500MG 052440-2.5G
101899-646EA 364.62 USD
101899-646 101899-648
N-Cyclohexyl-N-ethyl-2-piperidinecarboxamide hydrochloride
N-Cyclohexyl-N-ethyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₇ClN₂O
MDL Number: MFCD13562192
CAS Number: 1236263-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR