N-Benzyl-N-methyl-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052439-500MG
052439-2.5G
101899-642EA
364.62
USD
101899-642
101899-644
N-Benzyl-N-methyl-2-piperidinecarboxamide hydrochloride
N-Benzyl-N-methyl-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁ClN₂O |
MDL Number:
MFCD13562191 CAS Number: 205993-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...