N,N-Dipropyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052436-500MG 052436-2.5G
101899-630EA 364.62 USD
101899-630 101899-632
N,N-Dipropyl-2-piperidinecarboxamide hydrochloride
N,N-Dipropyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₅ClN₂O
MDL Number: MFCD13562188
CAS Number: 1236254-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR