(4-Hydroxy-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052435-500MG
052435-2.5G
101899-626EA
364.62
USD
101899-626
101899-628
(4-Hydroxy-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
(4-Hydroxy-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₁ClN₂O₂ MW: 248.76 g/mol |
MDL Number:
MFCD13562187 CAS Number: 1236256-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...