4-(2-Piperidinylcarbonyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052434-500MG 052434-2.5G
101899-622EA 364.62 USD
101899-622 101899-624
4-(2-Piperidinylcarbonyl)-2-piperazinone hydrochloride
4-(2-Piperidinylcarbonyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClN₃O₂
MDL Number: MFCD13562186
CAS Number: 1236262-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR