(4-Benzyl-1-piperazinyl)(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052432-500MG 052432-2.5G
101899-614EA 364.62 USD
101899-614 101899-616
(4-Benzyl-1-piperazinyl)(2-piperidinyl)methanone hydrochloride
(4-Benzyl-1-piperazinyl)(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₆ClN₃O
MDL Number: MFCD13562184
CAS Number: 1236266-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR