(4-Methyl-1-piperazinyl)(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052431-500MG 052431-2.5G
101899-610EA 364.62 USD
101899-610 101899-612
(4-Methyl-1-piperazinyl)(2-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperazinyl)(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClN₃O
MW: 247.77 g/mol
MDL Number: MFCD13562183
CAS Number: 1246172-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR