(4-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

Synonyms: 4-Methyl-1-(piperidin-2-ylcarbonyl)piperidine hydrochloride

052430-500MG 052430-2.5G
101899-606EA 364.62 USD
101899-606 101899-608
(4-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
(4-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MW: 246.78 g/mol
MDL Number: MFCD06407988
CAS Number: 690634-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR