(3-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052429-500MG 052429-2.5G
101899-602EA 364.62 USD
101899-602 101899-604
(3-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
(3-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MW: 246.78 g/mol
MDL Number: MFCD13562182
CAS Number: 1236263-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR