(2-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052428-500MG 052428-2.5G
101899-598EA 364.62 USD
101899-598 101899-600
(2-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride
(2-Methyl-1-piperidinyl)(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MW: 246.78 g/mol
MDL Number: MFCD13562181
CAS Number: 1236262-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR