N-Ethyl-N-(2-hydroxyethyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052426-500MG 052426-2.5G
101899-590EA 364.62 USD
101899-590 101899-592
N-Ethyl-N-(2-hydroxyethyl)-2-piperidinecarboxamide hydrochloride
N-Ethyl-N-(2-hydroxyethyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD13562179
CAS Number: 1236259-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR