N-Butyl-N-methyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052425-500MG 052425-2.5G
101899-586EA 364.62 USD
101899-586 101899-588
N-Butyl-N-methyl-2-piperidinecarboxamide hydrochloride
N-Butyl-N-methyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD13562178
CAS Number: 1246172-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR