(3-Hydroxy-1-pyrrolidinyl)(2-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052424-500MG 052424-2.5G
101899-582EA 364.62 USD
101899-582 101899-584
(3-Hydroxy-1-pyrrolidinyl)(2-piperidinyl)methanone hydrochloride
(3-Hydroxy-1-pyrrolidinyl)(2-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₂O₂
MDL Number: MFCD13562177
CAS Number: 1236261-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR